Gene Transcripts

Gene Symbol
Morc2b
Entrez Gene ID
299555
Gene Name
microrchidia 2B
Gene Aliases
RGD1564020
Chromosome Location
Chr.7: 15479754 - 15483834 on Build Rnor_6.0
Uni Gene
Rn.124768
Interrogated Sequence Translated Protein
RefSeq NM_001106764.1 NP_001100234.1

View Full Product Details