Gene Transcripts
- Gene Symbol
- LINC00472
- Entrez Gene ID
- 79940
- Gene Name
- long intergenic non-protein coding RNA 472
- Gene Aliases
- C6orf155
- Chromosome Location
- Chr. 6: 71407864 - 71420745 on Build GRCh38
- UniGene ID
- Hs.368337
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BF515823.1 |
- |
3 |
338 |
|
Chr. 6 - 71416460 - 71420266 [-] |
GenBankmRNA |
BF515614.1 |
- |
3 |
287 |
|
Chr. 6 - 71416410 - 71420266 [-] |
View Full Product Details