Gene Transcripts

Gene Symbol
LINC00472
Entrez Gene ID
79940
Gene Name
long intergenic non-protein coding RNA 472
Gene Aliases
C6orf155
Chromosome Location
Chr. 6: 71407864 - 71420745 on Build GRCh38
UniGene ID
Hs.368337
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026807.2 4 2644
NR_121612.1 4 2695
NR_121613.1 3 2629
NR_121614.1 3 2578
GenBank mRNA AK023251.1 - 4 2642

View Full Product Details