Gene Transcripts
- Gene Symbol
- LOC286359
- Entrez Gene ID
- 286359
- Gene Name
- uncharacterized LOC286359
- Gene Aliases
- Chromosome Location
- Chr. 9: 97390836 - 97396691 on Build GRCh38
- UniGene ID
- Hs.132262
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026847.1 |
|
6 |
904 |
|
GenBank mRNA |
AK097847.1 |
- |
1 |
1316 |
|
AL833510.1 |
- |
4 |
2345 |
|
View Full Product Details