Gene Transcripts
- Gene Symbol
- ZNF827
- Entrez Gene ID
- 152485
- Gene Name
- zinc finger protein 827
- Gene Aliases
- Chromosome Location
- Chr. 4: 145757627 - 145939008 on Build GRCh38
- UniGene ID
- Hs.133916
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG028101.1 |
- |
|
451 |
|
Chr. - - [-] |
GenBankmRNA |
CB242507.1 |
- |
1 |
594 |
|
Chr. 4 - 145757992 - 145758603 [-] |
GenBankmRNA |
BF680054.1 |
- |
1 |
473 |
|
Chr. 4 - 145757740 - 145758488 [-] |
View Full Product Details