Gene Transcripts
- Gene Symbol
- 4933424G06Rik
- Entrez Gene ID
- 71166
- Gene Name
- RIKEN cDNA 4933424G06 gene
- Gene Aliases
- 1700104I15Rik
- Chromosome Location
- Chr. 1: 36714538 - 36757960 on Build GRCm38
- UniGene ID
- Mm.157049
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK007119.1 |
- |
1 |
319 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY707198.1 |
- |
1 |
316 |
|
Chr. 1 - 36714549 - 36714938 [+] |
View Full Product Details