Gene Transcripts
- Gene Symbol
- Ccr9
- Entrez Gene ID
- 12769
- Gene Name
- chemokine (C-C motif) receptor 9
- Gene Aliases
- A130091K22Rik, Cmkbr10, GPR-9-6
- Chromosome Location
- Chr. 9: 123767211 - 123783457 on Build GRCm38
- UniGene ID
- Mm.440604
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK038272.1 |
- |
1 |
4104 |
|
AK162996.1 |
- |
1 |
4245 |
|
View Full Product Details