Gene Transcripts
- Gene Symbol
- LINC00472
- Entrez Gene ID
- 79940
- Gene Name
- long intergenic non-protein coding RNA 472
- Gene Aliases
- C6orf155
- Chromosome Location
- Chr. 6: 71407864 - 71420745 on Build GRCh38
- UniGene ID
- Hs.368337
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000426635.6 |
- |
4 |
992 |
|
Chr. 6 - 71414445 - 71420769 [-] |
GENCODE |
ENST00000602878.5 |
- |
3 |
469 |
|
Chr. 6 - 71416163 - 71420261 [-] |
GENCODE |
ENST00000421704.5 |
- |
3 |
424 |
|
Chr. 6 - 71416384 - 71420267 [-] |
View Full Product Details