Gene Transcripts
- Gene Symbol
- 8430436N08Rik
- Entrez Gene ID
- 71510
- Gene Name
- RIKEN cDNA 8430436N08 gene
- Gene Aliases
- Chromosome Location
- Chr. 4: 7560688 - 7573801 on Build GRCm38
- UniGene ID
- Mm.410775
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_040645.1 |
|
3 |
389 |
|
GenBank mRNA |
AK018465.1 |
- |
3 |
390 |
|
View Full Product Details