Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_038344.1 | 5 | 795 | ||
NR_038345.1 | 4 | 623 | |||
NR_038346.1 | 4 | 606 | |||
GenBank mRNA | AB371456.1 | - | 4 | 469 | |
BI868593.1 | - | 4 | 405 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB543548.1 | - | 2 | 213 | Chr. 7 - 79458936 - 79471194 [+] |