Gene Transcripts

Gene Symbol
Vmn1r82
Entrez Gene ID
171268
Gene Name
vomeronasal 1 receptor 82
Gene Aliases
V1rg12
Chromosome Location
Chr. 7: 12304805 - 12305719 on Build GRCm38
UniGene ID
Mm.377220
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134234.1 NP_598995.1 1 108
GenBank mRNA BC109325.2 - 1 108

View Full Product Details