Gene Transcripts
- Gene Symbol
- Ang4
- Entrez Gene ID
- 219033
- Gene Name
- angiogenin, ribonuclease A family, member 4
- Gene Aliases
- Raa4, Rnase5d
- Chromosome Location
- Chr. 14: 51763892 - 51773590 on Build GRCm38
- UniGene ID
- Mm.343899
- Species
- Mus musculus
- Gene Symbol
- Ang5
- Entrez Gene ID
- 503844
- Gene Name
- angiogenin, ribonuclease A family, member 5
- Gene Aliases
- Chromosome Location
- Chr. 14: 43957069 - 43963080 on Build GRCm38
- UniGene ID
- Mm.24663
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007448.3 |
NP_031474.2
|
2 |
472 |
|
View Full Product Details