Gene Transcripts

Gene Symbol
B3gnt8
Entrez Gene ID
232984
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 8
Gene Aliases
B3galt7, B7galt7, BC025206
Chromosome Location
Chr. 7: 25627587 - 25629492 on Build GRCm38
UniGene ID
Mm.192369
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001036740.2 NP_001031817.1 2 761
NM_146184.4 NP_666296.1 1 1153
GenBank mRNA AK042388.1 - 1 1136
AK154220.1 - 748
AK154901.1 - 2 717
BC025206.1 - 2 747 5037295

View Full Product Details