Gene Transcripts

Gene Symbol
Zfp759
Entrez Gene ID
268670
Gene Name
zinc finger protein 759
Gene Aliases
BC028265, Rslcan-8, Rslcan8
Chromosome Location
Chr. 13: 67125128 - 67142404 on Build GRCm38
UniGene ID
Mm.56274
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172392.3 NP_765980.3 4 1107
GenBank mRNA AK030126.1 - 4 1110
BC028265.1 - 4 1091 5365910

View Full Product Details