Gene Transcripts
- Gene Symbol
- 4930549O18Rik
- Entrez Gene ID
- 75335
- Gene Name
- RIKEN cDNA 4930549O18 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.389776
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK016081.1 |
- |
1, 2 |
205 |
|
- Gene Symbol
- 4930431C11Rik
- Entrez Gene ID
- 74641
- Gene Name
- RIKEN cDNA 4930431C11 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.416922
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK015261.1 |
- |
1, 2 |
208 |
|
View Full Product Details