Gene Transcripts
- Gene Symbol
- RPS16
- Entrez Gene ID
- 6217
- Gene Name
- ribosomal protein S16
- Gene Aliases
- S16
- Chromosome Location
- Chr. 19: 39433137 - 39436020 on Build GRCh38
- UniGene ID
- Hs.397609
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001020.5 |
NP_001011.1
|
3, 4 |
372 |
|
NM_001321111.1 |
NP_001308040.1
|
3, 4 |
321 |
|
GenBank mRNA |
AK055869.1 |
- |
2, 3 |
2145 |
|
AK315498.1 |
- |
3, 4 |
300 |
|
BC004324.2 |
- |
3, 4 |
298 |
3628799 |
BC007977.2 |
- |
3, 4 |
283 |
4310307 |
BC024970.1 |
- |
1 |
268 |
|
BC095429.1 |
- |
3, 4 |
242 |
|
BQ421722.1 |
- |
3, 4 |
330 |
|
CB107158.1 |
- |
3, 4 |
350 |
|
M60854.1 |
- |
3, 4 |
284 |
|
- Gene Symbol
- ZNF90
- Entrez Gene ID
- 7643
- Gene Name
- zinc finger protein 90
- Gene Aliases
- HTF9
- Chromosome Location
- Chr. 19: 20077994 - 20127032 on Build GRCh38
- UniGene ID
- Hs.55304
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK298173.1 |
- |
3 |
518 |
|
View Full Product Details