Gene Transcripts

Gene Symbol
Csdc2
Entrez Gene ID
266600
Gene Name
cold shock domain containing C2, RNA binding
Gene Aliases
Pippin
Chromosome Location
Chr. 7: 123168811 - 123183336 on Build Rnor_6.0
UniGene ID
Rn.1171
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170542.1 NP_001164013.1 1 165
GenBank mRNA CO405729.1 - 1 165
X89962.1 - 1 64

View Full Product Details