Gene Transcripts
- Gene Symbol
- Olr1385
- Entrez Gene ID
- 287236
- Gene Name
- olfactory receptor 1385
- Gene Aliases
- ratchr10-34281870-34280923_ORF
- Chromosome Location
- Chr. 10: 34360265 - 34361212 on Build Rnor_6.0
- UniGene ID
- Rn.142180
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_214833.1 |
NP_999998.1
|
1 |
457 |
|
- Gene Symbol
- Olr1383
- Entrez Gene ID
- 287232
- Gene Name
- olfactory receptor 1383
- Gene Aliases
- ratchr10-34221853-34220906_ORF
- Chromosome Location
- Chr. 10: 34300250 - 34301197 on Build Rnor_6.0
- UniGene ID
- Rn.142182
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_214832.1 |
NP_999997.1
|
1 |
457 |
|
View Full Product Details