Gene Transcripts

Gene Symbol
Pex19
Entrez Gene ID
289233
Gene Name
peroxisomal biogenesis factor 19
Gene Aliases
PxF
Chromosome Location
Chr. 13: 90514324 - 90530825 on Build Rnor_6.0
UniGene ID
Rn.203104
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134777.1 NP_001128249.1 7, 8 847

View Full Product Details