Gene Transcripts

Gene Symbol
Phgdh
Entrez Gene ID
58835
Gene Name
phosphoglycerate dehydrogenase
Gene Aliases
Chromosome Location
Chr. 2: 200484245 - 200513564 on Build Rnor_6.0
UniGene ID
Rn.6872
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031620.1 NP_113808.1 7, 8 755
XM_006233010.1 XP_006233072.1 6, 7 778
GenBank mRNA BC086327.1 - 6, 7 783
X97772.1 - 6, 7 714

View Full Product Details