Gene Transcripts

Gene Symbol
Cass4
Entrez Gene ID
296409
Gene Name
Cas scaffolding protein family member 4
Gene Aliases
RGD1310643
Chromosome Location
Chr. 3: 170398584 - 170438381 on Build Rnor_6.0
UniGene ID
Rn.228215
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191744.1 NP_001178673.1 5 1772
XM_006235679.2 XP_006235741.1 5 2681

View Full Product Details