Gene Transcripts

Gene Symbol
Klhdc2
Entrez Gene ID
299113
Gene Name
kelch domain containing 2
Gene Aliases
Chromosome Location
Chr. 6: 91595647 - 91610980 on Build Rnor_6.0
UniGene ID
Rn.129179
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034133.1 NP_001029305.1 2 383
GenBank mRNA BC105756.1 - 2 383

View Full Product Details