Gene Transcripts

Gene Symbol
Plekhg4
Entrez Gene ID
307796
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 4
Gene Aliases
RGD1310790
Chromosome Location
Chr. 19: 37330568 - 37345046 on Build Rnor_6.0
UniGene ID
Rn.110336
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006255483.2 XP_006255545.1 13 2509

View Full Product Details