Gene Transcripts
- Gene Symbol
- Adamtsl2
- Entrez Gene ID
- 311827
- Gene Name
- ADAMTS-like 2
- Gene Aliases
- RGD1305459
- Chromosome Location
- Chr. 3: 5624474 - 5654890 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_231125.7 |
XP_231125.3
|
9 |
1068 |
|
View Full Product Details