Gene Transcripts

Gene Symbol
Hscb
Entrez Gene ID
360826
Gene Name
HscB mitochondrial iron-sulfur cluster co-chaperone
Gene Aliases
RGD1311005
Chromosome Location
Chr. 12: 51878267 - 51888645 on Build Rnor_6.0
UniGene ID
Rn.65800
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108340.1 NP_001101810.1 5 613
XM_008769340.1 XP_008767562.1 6 792

View Full Product Details