Gene Transcripts

Gene Symbol
Ndst4
Entrez Gene ID
362035
Gene Name
N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 4
Gene Aliases
Chromosome Location
Chr. 2: 229195261 - 229496732 on Build Rnor_6.0
UniGene ID
Rn.162619
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191849.1 NP_001178778.1 8 1874
XM_006233274.2 XP_006233336.1 9 2195

View Full Product Details