Gene Transcripts

Gene Symbol
Olr145
Entrez Gene ID
365331
Gene Name
olfactory receptor 145
Gene Aliases
Chromosome Location
Chr. 1: 169269385 - 169270329 on Build Rnor_6.0
UniGene ID
Rn.141853
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000545.1 NP_001000545.1 1 486

View Full Product Details