Gene Transcripts

Gene Symbol
LOC103691702
Entrez Gene ID
103691702
Gene Name
60S ribosomal protein L29-like
Gene Aliases
Chromosome Location
Chr. 2: 221571856 - 221572200 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008761486.1 XP_008759708.1 1 71

View Full Product Details