Gene Transcripts

Gene Symbol
Syf2
Entrez Gene ID
170933
Gene Name
SYF2 pre-mRNA-splicing factor
Gene Aliases
Gcipip, P29
Chromosome Location
Chr. 5: 153269638 - 153277562 on Build Rnor_6.0
UniGene ID
Rn.162117
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133417.1 NP_596908.1 4, 5 402
GenBank mRNA AF366369.1 - 4, 5 402
BC097289.1 - 4, 5 410 7445525
FQ234066.1 - 2, 3 229

View Full Product Details