Gene Transcripts

Gene Symbol
HEATR5A
Entrez Gene ID
25938
Gene Name
HEAT repeat containing 5A
Gene Aliases
C14orf125
Chromosome Location
Chr. 14: 31291788 - 31420582 on Build GRCh38
UniGene ID
Hs.744979
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015473.3 NP_056288.2 19 2998
GenBank mRNA AB037737.1 - 10 1446
AK092400.1 - 12 1740
BC045764.1 - 13 2011
BC063867.1 - 1 22
CR749444.1 - 12 1696

View Full Product Details