Gene Transcripts

Gene Symbol
Gltscr1
Entrez Gene ID
292622
Gene Name
glioma tumor suppressor candidate region gene 1
Gene Aliases
Chromosome Location
Chr. 1: 77869020 - 77943836 on Build Rnor_6.0
UniGene ID
Rn.167279
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106226.2 NP_001099696.2 8, 9 3210
XM_006228343.2 XP_006228405.1 10, 11 3557
XM_006228344.2 XP_006228406.1 9, 10 3491
XM_006228345.2 XP_006228407.1 9, 10 3380

View Full Product Details