Gene Transcripts

Gene Symbol
NCAPD3
Entrez Gene ID
23310
Gene Name
non-SMC condensin II complex subunit D3
Gene Aliases
CAP-D3, CAPD3, hCAP-D3, hHCP-6, hcp-6
Chromosome Location
Chr. 11: 134152442 - 134225443 on Build GRCh38
UniGene ID
Hs.438550
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015261.2 NP_056076.1 26 3942
XM_011542719.2 XP_011541021.1 25 3203
GenBank mRNA AK025667.1 - 10 1233
AK124878.1 - 26 3868
BC011408.1 - 4 517
BC098398.1 - 26 3401 30528692
BC144524.1 - 27 3767
D29954.1 - 26 3364

View Full Product Details