Gene Transcripts

Gene Symbol
Rpl27
Entrez Gene ID
64306
Gene Name
ribosomal protein L27
Gene Aliases
Chromosome Location
Chr. 10: 89352864 - 89356563 on Build Rnor_6.0
UniGene ID
Rn.1254
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022514.1 NP_071959.1 4 396
GenBank mRNA BC058474.1 - 5 426
BC091566.1 - 5 394 7388530
FQ210011.1 - 5 412
FQ214023.1 - 5 416
FQ217713.1 - 4 336
FQ221292.1 - 5 421
FQ221643.1 - 5 413
FQ221684.1 - 5 414
FQ221938.1 - 5 427
FQ222018.1 - 5 416
FQ222606.1 - 5 417
FQ222991.1 - 5 418
FQ223942.1 - 4 345
FQ224664.1 - 5 414
FQ228505.1 - 5 413
FQ228603.1 - 5 414
FQ228786.1 - 5 413
X07424.1 - 4 396

View Full Product Details