Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014053.3 | NP_054772.1 | 3 | 1163 | |
XM_011509446.2 | XP_011507748.1 | 3 | 1139 | ||
XM_011509447.2 | XP_011507749.1 | 5 | 1381 | ||
GenBank mRNA | AF118637.1 | - | 3 | 1163 | |
AK001419.1 | - | 3 | 340 | ||
AK314481.1 | - | 3 | 1139 | ||
BC048312.1 | - | 3 | 1101 | 4417876 |