Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001712.2 | NP_001001712.2 | 4 | 503 | |
GenBank mRNA | AK091788.1 | - | 2 | 204 | |
AY301271.1 | - | 3 | 408 | ||
BC040963.1 | - | 4 | 1748 | ||
BC064596.1 | - | 2 | 176 | ||
BC133045.1 | - | 4 | 503 | 40147067 |