Gene Transcripts

Gene Symbol
OR2W5
Entrez Gene ID
441932
Gene Name
olfactory receptor family 2 subfamily W member 5 (gene/pseudogene)
Gene Aliases
OR2W5P, OST722
Chromosome Location
Chr. 1: 247491068 - 247492409 on Build GRCh38
UniGene ID
Hs.591526
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004698.2 NP_001004698.1 1 827
GenBank mRNA BC137203.1 - 1 827
BC137204.1 - 1 827 9021201

View Full Product Details