Gene Transcripts

Gene Symbol
Sfmbt1
Entrez Gene ID
54650
Gene Name
Scm-like with four mbt domains 1
Gene Aliases
4930442N21Rik, 9330180L21Rik, AA536974, Sfmbt, Smr
Chromosome Location
Chr. 14: 30714473 - 30822721 on Build GRCm38
UniGene ID
Mm.294662
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166531.1 NP_001160003.1 15, 16 1749
NM_001166532.1 NP_001160004.1 14, 15 1843
NM_019460.2 NP_062333.1 14, 15 1700
GenBank mRNA AB032165.1 - 13, 14 1545
AK142330.1 - 14, 15 1728
AK154826.1 - 14, 15 1845
AK160573.1 - 15, 16 1749
BC018217.1 - 14, 15 1711
BC036972.1 - 14, 15 1770
BC066188.1 - 14, 15 1653 30476111

View Full Product Details