Gene Transcripts

Gene Symbol
Rbm8a
Entrez Gene ID
60365
Gene Name
RNA binding motif protein 8a
Gene Aliases
2310057C03Rik, AA673428, Rbm8
Chromosome Location
Chr. 3: 96629928 - 96633791 on Build GRCm38
UniGene ID
Mm.261972
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102407.1 NP_001095877.1 6 1208
NM_025875.2 NP_080151.2 6 1205
GenBank mRNA AK009953.1 - 6 1199

View Full Product Details