Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028299.1 | NP_082575.1 | 6 | 763 | |
GenBank mRNA | AK012304.1 | - | 6 | 736 | |
AK150453.1 | - | 6 | 749 | ||
AK151754.1 | - | 5 | 580 | ||
AK157260.1 | - | 6 | 749 | ||
BC042740.1 | - | 6 | 763 | 5024951 |