Gene Transcripts

Gene Symbol
4930402K13Rik
Entrez Gene ID
75775
Gene Name
RIKEN cDNA 4930402K13 gene
Gene Aliases
Chromosome Location
Chr. X: 9104562 - 9106342 on Build GRCm38
UniGene ID
Mm.50374
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270700.1 NP_001257629.1 1 1163
GenBank mRNA AK015052.1 - 1 1164

View Full Product Details