Gene Transcripts

Gene Symbol
Crisp4
Entrez Gene ID
78081
Gene Name
cysteine-rich secretory protein 4
Gene Aliases
9230112K08Rik
Chromosome Location
Chr. 1: 18114905 - 18145967 on Build GRCm38
UniGene ID
Mm.319913
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030033.1 NP_084309.1 6 563
GenBank mRNA AK020340.1 - 7 624
AY705449.1 - 6 563
DQ200908.1 - 8 740

View Full Product Details