Gene Transcripts

Gene Symbol
Plekha3
Entrez Gene ID
83435
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 3
Gene Aliases
FAPP1
Chromosome Location
Chr. 2: 76675310 - 76697335 on Build GRCm38
UniGene ID
Mm.41636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031256.3 NP_112546.1 4 678
GenBank mRNA AF286163.1 - 4 467
BC012283.1 - 4 418
BC031110.1 - 5 486 4981486

View Full Product Details