Search
Search

Igbp1| Gene Symbol: Igbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_008784.3 | NP_032810.4 | ||||
| GenBank mRNA | AA213189.1 | - | ||||
| AK017166.1 | - | |||||
| AK032709.1 | - | |||||
| AK141470.1 | - | |||||
| AK156610.1 | - | |||||
| AK186364.1 | - | |||||
| AK210875.1 | - | |||||
| AK218313.1 | - | |||||
| BC056186.1 | - | |||||
| BC089567.1 | - | 30297519 | ||||
| BU520087.1 | - | |||||
| BY018773.1 | - | |||||
| CF585770.1 | - | |||||
| CX203560.1 | - | |||||
| CX567459.1 | - | |||||
| DV654442.1 | - | |||||
| DV655610.1 | - | |||||
| L31652.1 | - | |||||