Search
Search

Rnf112| Gene Symbol: Rnf112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | IMAGE Clone ID | ||||
| RefSeq | NM_001291024.1 | NP_001277953.1 | ||||
| NM_009548.3 | NP_033574.2 | |||||
| GenBank mRNA | AB013097.1 | - | ||||
| AK131651.1 | - | |||||
| AK165038.1 | - | |||||
| BC013139.1 | - | 4500921 | ||||
| BC037118.1 | - | 4506141 | ||||
| BY599807.1 | - | |||||
| CJ148450.1 | - | |||||