Search
Search

| Assay Name: | mmu-miR-466a-3p |
|---|---|
| miRBase Accession Number: | MI0002401 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAUACAUACACGCACACAUAAGA |
| Species: | Mouse |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Gene Family ID: | MIPF0000208, mir-466 |
Mature miRNA Sequence: | UAUACAUACACGCACACAUAAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-466a-3p | MIMAT0002107 |
| Mouse | mmu-miR-466e-3p | MIMAT0004880 |