Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › PM11353
          Assay Name: hsa-miR-363-5p
          miRBase Accession Number: MI0000764
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 134169378 - 134169452 [-] on Build GRCh38
          Mature miRNA Sequence: CGGGUGGAUCACGAUGCAAUUU
          Species: Human, Rat, Goat, Ophiophagus hannah, Ornithorhynchus anatinus, Xenopus laevis, Xenopus tropicalis
          Product Type: Ambion® Pre-miR™ miRNA Precursor
          Assay ID PM11353
          Size
          Availability Made To Order
          Catalog # AM17100
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000138, mir-363

          Mature miRNA Sequence:

          CGGGUGGAUCACGAUGCAAUUU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-363-5p MIMAT0003385
          Rat rno-miR-363-5p MIMAT0003209
          Goat chi-miR-363-5p MIMAT0036175
          Ophiophagus hannah oha-miR-363-5p MIMAT0036912
          Ornithorhynchus anatinus oan-miR-363-5p MIMAT0006859
          Xenopus laevis xla-miR-363 MIMAT0011147
          View More Rows
          Xenopus tropicalis xtr-miR-363-5p MIMAT0003701

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-363
          Stem-loop Accession # MI0000764
          Stem-loop Sequence
          UGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUGAGUAUCAUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACC
          Chromosome Location Chr. X - 134169378 - 134169452 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0003385
          miRBase ID: hsa-miR-363-5p
          Mature miRNA Sequence: CGGGUGGAUCACGAUGCAAUUU
          Chromosome Location: Chr. X - 134169378 - 134169452 [-] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-363
          Stem-loop Accession # MI0003553
          Stem-loop Sequence
          UUUUGCUGUUAUCGGGUGGAUCACGAUGCAAUUUUGAUUAGAGUAAUGGGAGAGAAAUUGCACGGUAUCCAUCUGUAAACCGCAGGA
          Chromosome Location Chr. X - 140117098 - 140117184 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003209
          miRBase ID: rno-miR-363-5p
          Mature miRNA Sequence: CGGGUGGAUCACGAUGCAAUUU
          Chromosome Location: Chr. X - 140117098 - 140117184 [-] on Build Rnor_6.0

          Goat

          Stem-loop ID chi-mir-363
          Stem-loop Accession # MI0030762
          Stem-loop Sequence
          UUGUAAAAUGACCUGUUUUGCUGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUAUAAUAGGAGAAAAAUUGCACGGUAUCCAUCUGCGAACCGCAGGACCCUUGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036175
          miRBase ID: chi-miR-363-5p
          Mature miRNA Sequence: CGGGUGGAUCACGAUGCAAUUU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-363
          Stem-loop Accession # MI0031463
          Stem-loop Sequence
          GUUGCUGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGAUUAGCAGGAAGGAAAUUGCACGGUAUCCAUCUGUAAACCGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036912
          miRBase ID: oha-miR-363-5p
          Mature miRNA Sequence: CGGGUGGAUCACGAUGCAAUUU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-363
          Stem-loop Accession # MI0006698
          Stem-loop Sequence
          GUUUUGCUGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUUUAGUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACCGCAGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006859
          miRBase ID: oan-miR-363-5p
          Mature miRNA Sequence: CGGGUGGAUCACGAUGCAAUUU
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-363
          Stem-loop Accession # MI0008385
          Stem-loop Sequence
          UUUUGUUGUUCGCGGGUGGAUCACGAUGCAAUUUUAUUUAAGUUUGGUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACCGCAGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0011147
          miRBase ID: xla-miR-363
          Mature miRNA Sequence: CGGGUGGAUCACGAUGCAAUUU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-363
          Stem-loop Accession # MI0004955
          Stem-loop Sequence
          UUUUGUUGUUCGCGGGUGGAUCACGAUGCAAUUUUAUUUAGUUUGGUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACCGCAGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003701
          miRBase ID: xtr-miR-363-5p
          Mature miRNA Sequence: CGGGUGGAUCACGAUGCAAUUU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465449_pri
          TaqMan™ Pri-miRNA Assay : Hs03295977_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11353
          TaqMan™ MicroRNA Assay : 001283
          mirVana® miRNA inhibitor : MH11353
          mirVana® miRNA mimic : MC11353
          TaqMan™ Advanced miRNA Assay : rno478840_mir
          TaqMan™ Advanced miRNA Assay : 478840_mir


          miRBase Alias:

          Human: hsa-miR-363* (v17)
          Rat: rno-miR-363* (v18)
          Ochrobactrum anthropi: oan-miR-363* (v18)

          Back To Top

          Related Products

          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline