Product References
FACS and immunomagnetic isolation of early erythroid progenitor cells from mouse fetal liver.
STAR protocols
Braun TW,Kuoch MK,Khandros E,Li H
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Fri Mar 18 00:00:00 EDT 2022
IL-22 initiates an IL-18-dependent epithelial response circuit to enforce intestinal host defence.
Nature communications
Chiang HY,Lu HH,Sudhakar JN,Chen YW,Shih NS,Weng YT,Shui JW
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Tue Feb 15 00:00:00 EST 2022
Ependymoma associated protein Zfta is expressed in immature ependymal cells but is not essential for ependymal development in mice.
Scientific reports
Herranz-Pérez V,Nakatani J,Ishii M,Katada T,García-Verdugo JM,Ohata S
11-0242-82 was used in Immunohistochemistry to explore the expression profile and potential function of ZFTA (zinc finger translocation associated) in the development of the ependymal (E1) cells of the brain.
Thu Jan 27 00:00:00 EST 2022
Cytosolic Nuclear Sensor Dhx9 Controls Medullary Thymic Epithelial Cell Differentiation by p53-Mediated Pathways.
Frontiers in immunology
Dong X,Zhang J,Zhang Q,Liang Z,Xu Y,Zhao Y,Zhang B
11-0242-82 was used in Flow cytometry/Cell sorting to reveal that Dhx9, a cytosolic nuclear sensor recognizes viral DNA or RNA, playing an important role in mTEC development and function in mice.
Tue Jun 21 00:00:00 EDT 2022
Hormone-Responsive BMP Signaling Expands Myoepithelial Cell Lineages and Prevents Alveolar Precocity in Mammary Gland.
Frontiers in cell and developmental biology
Shao C,Lou P,Liu R,Bi X,Li G,Yang X,Sheng X,Xu J,Lv C,Yu Z
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Tue Aug 03 00:00:00 EDT 2021
Testosterone metabolites differentially regulate obesogenesis and fat distribution.
Molecular metabolism
Sebo ZL,Rodeheffer MS
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Mon Feb 01 00:00:00 EST 2021
Membrane expression of the estrogen receptor ERα is required for intercellular communications in the mammary epithelium.
Development (Cambridge, England)
Gagniac L,Rusidzé M,Boudou F,Cagnet S,Adlanmerini M,Jeannot P,Gaide N,Giton F,Besson A,Weyl A,Gourdy P,Raymond-Letron I,Arnal JF,Brisken C,Lenfant F
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Wed Mar 11 00:00:00 EDT 2020
Analysis of Myeloid Cells in Mouse Tissues with Flow Cytometry.
STAR protocols
Liu Z,Gu Y,Shin A,Zhang S,Ginhoux F
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Fri Jun 19 00:00:00 EDT 2020
Analysis of Myeloid Cells in Mouse Tissues with Flow Cytometry.
STAR protocols
Liu Z,Gu Y,Shin A,Zhang S,Ginhoux F
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Fri Jun 19 00:00:00 EDT 2020
Transcriptomic and proteomic signatures of stemness and differentiation in the colon crypt.
Communications biology
Habowski AN,Flesher JL,Bates JM,Tsai CF,Martin K,Zhao R,Ganesan AK,Edwards RA,Shi T,Wiley HS,Shi Y,Hertel KJ,Waterman ML
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Wed Aug 19 00:00:00 EDT 2020
An Nfil3-Zeb2-Id2 pathway imposes Irf8 enhancer switching during cDC1 development.
Nature immunology
Bagadia P,Huang X,Liu TT,Durai V,Grajales-Reyes GE,Nitschké M,Modrusan Z,Granja JM,Satpathy AT,Briseño CG,Gargaro M,Iwata A,Kim S,Chang HY,Shaw AS,Murphy TL,Murphy KM
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Sun Sep 01 00:00:00 EDT 2019
Lifelong n-3 Polyunsaturated Fatty Acid Exposure Modulates Size of Mammary Epithelial Cell Populations and Expression of Caveolae Resident Proteins in Fat-1 Mice.
Nutrients
Hillyer LM,Kang JX,Ma DWL
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Tue Oct 15 00:00:00 EDT 2019
A ZEB1/p53 signaling axis in stromal fibroblasts promotes mammary epithelial tumours.
Nature communications
Fu R,Han CF,Ni T,Di L,Liu LJ,Lv WC,Bi YR,Jiang N,He Y,Li HM,Wang S,Xie H,Chen BA,Wang XS,Weiss SJ,Lu T,Guo QL,Wu ZQ
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Fri Jul 19 00:00:00 EDT 2019
A ZEB1/p53 signaling axis in stromal fibroblasts promotes mammary epithelial tumours.
Nature communications
Fu R,Han CF,Ni T,Di L,Liu LJ,Lv WC,Bi YR,Jiang N,He Y,Li HM,Wang S,Xie H,Chen BA,Wang XS,Weiss SJ,Lu T,Guo QL,Wu ZQ
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Fri Jul 19 00:00:00 EDT 2019
Adipocyte hypertrophy and lipid dynamics underlie mammary gland remodeling after lactation.
Nature communications
Zwick RK,Rudolph MC,Shook BA,Holtrup B,Roth E,Lei V,Van Keymeulen A,Seewaldt V,Kwei S,Wysolmerski J,Rodeheffer MS,Horsley V
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Tue Sep 04 00:00:00 EDT 2018
Wnt Effector TCF4 Is Dispensable for Wnt Signaling in Human Cancer Cells.
Genes
Hrckulak D,Janeckova L,Lanikova L,Kriz V,Horazna M,Babosova O,Vojtechova M,Galuskova K,Sloncova E,Korinek V
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Sat Sep 01 00:00:00 EDT 2018
ATF3 Sustains IL-22-Induced STAT3 Phosphorylation to Maintain Mucosal Immunity Through Inhibiting Phosphatases.
Frontiers in immunology
Glal D,Sudhakar JN,Lu HH,Liu MC,Chiang HY,Liu YC,Cheng CF,Shui JW
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Thu Sep 26 00:00:00 EDT 2019
Neonatally imprinted stromal cell subsets induce tolerogenic dendritic cells in mesenteric lymph nodes.
Nature communications
Pezoldt J,Pasztoi M,Zou M,Wiechers C,Beckstette M,Thierry GR,Vafadarnejad E,Floess S,Arampatzi P,Buettner M,Schweer J,Fleissner D,Vital M,Pieper DH,Basic M,Dersch P,Strowig T,Hornef M,Bleich A,Bode U,Pabst O,Bajénoff M,Saliba AE,Huehn J
11-0242 was used in Flow cytometry/Cell sorting to study the function of the neonatal imprinting of stromal cells in mesenteric lymph nodes.
Tue Sep 25 00:00:00 EDT 2018
Single-Cell Analysis Identifies LY6D as a Marker Linking Castration-Resistant Prostate Luminal Cells to Prostate Progenitors and Cancer.
Cell reports
Barros-Silva JD,Linn DE,Steiner I,Guo G,Ali A,Pakula H,Ashton G,Peset I,Brown M,Clarke NW,Bronson RT,Yuan GC,Orkin SH,Li Z,Baena E
11-0242 was used in Flow cytometry/Cell sorting to analyse the molecular heterogeneity in basal and luminal prostate compartments.
Tue Dec 18 00:00:00 EST 2018
Stabilization of cytokine mRNAs in iNKT cells requires the serine-threonine kinase IRE1alpha.
Nature communications
Govindarajan S,Gaublomme D,Van der Cruyssen R,Verheugen E,Van Gassen S,Saeys Y,Tavernier S,Iwawaki T,Bloch Y,Savvides SN,Lambrecht BN,Janssens S,Elewaut D,Drennan MB
11-0242 was used in Flow cytometry/Cell sorting to examine the role of the IRE1α-dependent signalling cascade in coupling constitutive cytokine mRNA expression to the rapid induction of cytokine secretion and effector functions in activated iNKT cells.
Mon Dec 17 00:00:00 EST 2018
A Paradoxical Tumor-Suppressor Role for the Rac1 Exchange Factor Vav1 in T Cell Acute Lymphoblastic Leukemia.
Cancer cell
Robles-Valero J,Lorenzo-Martín LF,Menacho-Márquez M,Fernández-Pisonero I,Abad A,Camós M,Toribio ML,Espinosa L,Bigas A,Bustelo XR
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Mon Nov 13 00:00:00 EST 2017
Adipocyte Accumulation in the Bone Marrow during Obesity and Aging Impairs Stem Cell-Based Hematopoietic and Bone Regeneration.
Cell stem cell
Ambrosi TH,Scialdone A,Graja A,Gohlke S,Jank AM,Bocian C,Woelk L,Fan H,Logan DW,Schürmann A,Saraiva LR,Schulz TJ
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
Metabolic shifts in residual breast cancer drive tumor recurrence.
The Journal of clinical investigation
Havas KM,Milchevskaya V,Radic K,Alladin A,Kafkia E,Garcia M,Stolte J,Klaus B,Rotmensz N,Gibson TJ,Burwinkel B,Schneeweiss A,Pruneri G,Patil KR,Sotillo R,Jechlinger M
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Thu Jun 01 00:00:00 EDT 2017
CD24 induces changes to the surface receptors of B cell microvesicles with variable effects on their RNA and protein cargo.
Scientific reports
Ayre DC,Chute IC,Joy AP,Barnett DA,Hogan AM,Grüll MP,Peña-Castillo L,Lang AS,Lewis SM,Christian SL
11-0242 was used in Flow cytometry/Cell sorting to perform a systematic characterisation of B cell microvesicles released from WEHI-231 B lymphoma cells in response to CD24 stimulation.
Thu Aug 17 00:00:00 EDT 2017
CD24 induces changes to the surface receptors of B cell microvesicles with variable effects on their RNA and protein cargo.
Scientific reports
Ayre DC,Chute IC,Joy AP,Barnett DA,Hogan AM,Grüll MP,Peña-Castillo L,Lang AS,Lewis SM,Christian SL
11-0242 was used in Flow cytometry/Cell sorting to perform a systematic characterisation of B cell microvesicles released from WEHI-231 B lymphoma cells in response to CD24 stimulation.
Thu Aug 17 00:00:00 EDT 2017
Epithelia Use Butyrophilin-like Molecules to Shape Organ-Specific γδ T Cell Compartments.
Cell
Di Marco Barros R,Roberts NA,Dart RJ,Vantourout P,Jandke A,Nussbaumer O,Deban L,Cipolat S,Hart R,Iannitto ML,Laing A,Spencer-Dene B,East P,Gibbons D,Irving PM,Pereira P,Steinhoff U,Hayday A
11-0242 was used in Flow cytometry/Cell sorting to investigate the mechanism that shape organ-specific γδ T cell compartments, showing that Btnl1 expressed by murine enterocytes shapes the local γδ compartment, as does BTNL3 and BTNL8 expressed by human gut epithelial cells, suggesting a conserved mechanism shaped local T cell compartments.
Thu Sep 22 00:00:00 EDT 2016
Skin Adipocyte Stem Cell Self-Renewal Is Regulated by a PDGFA/AKT-Signaling Axis.
Cell stem cell
Rivera-Gonzalez GC,Shook BA,Andrae J,Holtrup B,Bollag K,Betsholtz C,Rodeheffer MS,Horsley V
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
Skin Adipocyte Stem Cell Self-Renewal Is Regulated by a PDGFA/AKT-Signaling Axis.
Cell stem cell
Rivera-Gonzalez GC,Shook BA,Andrae J,Holtrup B,Bollag K,Betsholtz C,Rodeheffer MS,Horsley V
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Thu Dec 01 00:00:00 EST 2016
NKT sublineage specification and survival requires the ubiquitin-modifying enzyme TNFAIP3/A20.
The Journal of experimental medicine
Drennan MB,Govindarajan S,Verheugen E,Coquet JM,Staal J,McGuire C,Taghon T,Leclercq G,Beyaert R,van Loo G,Lambrecht BN,Elewaut D
11-0242 was used in Flow cytometry/Cell sorting to show that negative regulation of TCR signalling during NKT development controls the differentiation and survival of NKT1 and NKT2 cells.
Mon Sep 19 00:00:00 EDT 2016
Casitas B-cell lymphoma (Cbl) proteins protect mammary epithelial cells from proteotoxicity of active c-Src accumulation.
Proceedings of the National Academy of Sciences of the United States of America
Mukhopadhyay C,Triplett A,Bargar T,Heckman C,Wagner KU,Naramura M
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Immunofluorescence
Tue Dec 20 00:00:00 EST 2016
Severe Changes in Thymic Microenvironment in a Chronic Experimental Model of Paracoccidioidomycosis.
PloS one
Alves da Costa T,Di Gangi R,Thomé R,Barreto Felisbino M,Pires Bonfanti A,Lumi Watanabe Ishikawa L,Sartori A,Burger E,Verinaud L
11024282 was used in flow cytometry to examine the thymic microenvironment using a model of paracoccidioidomycosis
Mon May 22 00:00:00 EDT 2017
Rapid depot-specific activation of adipocyte precursor cells at the onset of obesity.
Nature cell biology
Jeffery E,Church CD,Holtrup B,Colman L,Rodeheffer MS
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Wed Apr 01 00:00:00 EDT 2015
The immunosenescence-related gene Zizimin2 is associated with early bone marrow B cell development and marginal zone B cell formation.
Immunity & ageing : I & A
Matsuda T,Yanase S,Takaoka A,Maruyama M
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Mon Mar 02 00:00:00 EST 2015
Dynamic regulation of CD24 expression and release of CD24-containing microvesicles in immature B cells in response to CD24 engagement.
Immunology
Ayre DC,Elstner M,Smith NC,Moores ES,Hogan AM,Christian SL
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2015
Dynamic regulation of CD24 expression and release of CD24-containing microvesicles in immature B cells in response to CD24 engagement.
Immunology
Ayre DC,Elstner M,Smith NC,Moores ES,Hogan AM,Christian SL
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2015
Dynamic regulation of CD24 expression and release of CD24-containing microvesicles in immature B cells in response to CD24 engagement.
Immunology
Ayre DC,Elstner M,Smith NC,Moores ES,Hogan AM,Christian SL
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Thu Oct 01 00:00:00 EDT 2015
Does FACS perturb gene expression?
Cytometry. Part A : the journal of the International Society for Analytical Cytology
Richardson GM,Lannigan J,Macara IG
11-0242 was used in Flow cytometry/Cell sorting to examine the effects of fluorescence activated cell sorting separation on short-term transcriptional profiles.
Sun Feb 01 00:00:00 EST 2015
The Transcriptional Repressor ZNF503/Zeppo2 Promotes Mammary Epithelial Cell Proliferation and Enhances Cell Invasion.
The Journal of biological chemistry
Shahi P,Slorach EM,Wang CY,Chou J,Lu A,Ruderisch A,Werb Z
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Fri Feb 06 00:00:00 EST 2015
FGF ligands of the postnatal mammary stroma regulate distinct aspects of epithelial morphogenesis.
Development (Cambridge, England)
Zhang X,Martinez D,Koledova Z,Qiao G,Streuli CH,Lu P
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Mon Sep 01 00:00:00 EDT 2014
Rhof promotes murine marginal zone B cell development.
Nagoya journal of medical science
Kishimoto M,Matsuda T,Yanase S,Katsumi A,Suzuki N,Ikejiri M,Takagi A,Ikawa M,Kojima T,Kunishima S,Kiyoi H,Naoe T,Matsushita T,Maruyama M
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Fri Aug 01 00:00:00 EDT 2014
Prospective identification of functionally distinct stem cells and neurosphere-initiating cells in adult mouse forebrain.
eLife
Mich JK,Signer RA,Nakada D,Pineda A,Burgess RJ,Vue TY,Johnson JE,Morrison SJ
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Wed May 07 00:00:00 EDT 2014
Leukocyte population dynamics and detection of IL-9 as a major cytokine at the mouse fetal-maternal interface.
PloS one
Habbeddine M,Verbeke P,Karaz S,Bobé P,Kanellopoulos-Langevin C
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Wed Jun 10 00:00:00 EDT 2015
Characterization of the adipocyte cellular lineage in vivo.
Nature cell biology
Berry R,Rodeheffer MS
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Fri Mar 01 00:00:00 EST 2013
Evidence for a multipotent mammary progenitor with pregnancy-specific activity.
Breast cancer research : BCR
Kaanta AS,Virtanen C,Selfors LM,Brugge JS,Neel BG
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Mon Dec 08 00:00:00 EST 2014
FLT3-ITDs instruct a myeloid differentiation and transformation bias in lymphomyeloid multipotent progenitors.
Cell reports
Mead AJ,Kharazi S,Atkinson D,Macaulay I,Pecquet C,Loughran S,Lutteropp M,Woll P,Chowdhury O,Luc S,Buza-Vidas N,Ferry H,Clark SA,Goardon N,Vyas P,Constantinescu SN,Sitnicka E,Nerlov C,Jacobsen SE
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Thu Jun 27 00:00:00 EDT 2013
Lunatic fringe deficiency cooperates with the Met/Caveolin gene amplicon to induce basal-like breast cancer.
Cancer cell
Xu K,Usary J,Kousis PC,Prat A,Wang DY,Adams JR,Wang W,Loch AJ,Deng T,Zhao W,Cardiff RD,Yoon K,Gaiano N,Ling V,Beyene J,Zacksenhaus E,Gridley T,Leong WL,Guidos CJ,Perou CM,Egan SE
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Tue May 15 00:00:00 EDT 2012
Restricted expression of new guanine nucleotide exchange factor Zizimin2 in aged acquired immune system.
Nagoya journal of medical science
Jia Y,Sakabe I,Matsuda T,Hayakawa T,Maruyama M
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Wed Aug 01 00:00:00 EDT 2012
Stem cell marker prominin-1 regulates branching morphogenesis, but not regenerative capacity, in the mammary gland.
Developmental dynamics : an official publication of the American Association of Anatomists
Anderson LH,Boulanger CA,Smith GH,Carmeliet P,Watson CJ
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Tue Mar 01 00:00:00 EST 2011
A multifunctional 3D co-culture system for studies of mammary tissue morphogenesis and stem cell biology.
PloS one
Campbell JJ,Davidenko N,Caffarel MM,Cameron RE,Watson CJ
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Flow Cytometry
Mon Mar 05 00:00:00 EST 2012
Robust in vivo transduction of nervous system and neural stem cells by early gestational intra amniotic gene transfer using lentiviral vector.
Molecular therapy : the journal of the American Society of Gene Therapy
Stitelman DH,Endo M,Bora A,Muvarak N,Zoltick PW,Flake AW,Brazelton TR
Published figure using CD24 monoclonal antibody (Product # 11-0242-82) in Immunofluorescence
Wed Sep 01 00:00:00 EDT 2010
The adaptor molecule signaling lymphocytic activation molecule-associated protein (SAP) regulates IFN-gamma and IL-4 production in V alpha 14 transgenic NKT cells via effects on GATA-3 and T-bet expression.
Journal of immunology (Baltimore, Md. : 1950)
Cen O,Ueda A,Guzman L,Jain J,Bassiri H,Nichols KE,Stein PL
11-0242 was used in Flow cytometry/Cell sorting to study the mechanism by which signalling lymphocytic activation molecule-associated protein exerts control over natural killer T cell development and cytokine production.
Sun Feb 01 00:00:00 EST 2009
Selective expression rather than specific function of Txk and Itk regulate Th1 and Th2 responses.
Journal of immunology (Baltimore, Md. : 1950)
Sahu N,Venegas AM,Jankovic D,Mitzner W,Gomez-Rodriguez J,Cannons JL,Sommers C,Love P,Sher A,Schwartzberg PL,August A
11-0242 was used in Flow cytometry/Cell sorting to determine that Itk and Txk exert their effects on Th cell differentiation/function at the level of expression.
Sat Nov 01 00:00:00 EDT 2008
An MHC-linked locus modulates thymic differentiation of CD4+CD25+Foxp3+ regulatory T lymphocytes.
International immunology
Tellier J,van Meerwijk JP,Romagnoli P
11-0242 was used in Flow cytometry/Cell sorting to investigate factors that affect the levels of Tregs in the thymus.
Wed Nov 01 00:00:00 EST 2006