Product References
Rapid increase in transferrin receptor recycling promotes adhesion during T cell activation.
BMC biology
Rossatti P,Redpath GMI,Ziegler L,Samson GPB,Clamagirand CD,Legler DF,Rossy J
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Wed Aug 24 00:00:00 EDT 2022
A patient-designed tissue-engineered model of the infiltrative glioblastoma microenvironment.
NPJ precision oncology
Cornelison RC,Yuan JX,Tate KM,Petrosky A,Beeghly GF,Bloomfield M,Schwager SC,Berr AL,Stine CA,Cimini D,Bafakih FF,Mandell JW,Purow BW,Horton BJ,Munson JM
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Fri Jul 29 00:00:00 EDT 2022
Decoding the pathogenesis of Diamond-Blackfan anemia using single-cell RNA-seq.
Cell discovery
Wang B,Wang C,Wan Y,Gao J,Ma Y,Zhang Y,Tong J,Zhang Y,Liu J,Chang L,Xu C,Shen B,Chen Y,Jiang E,Kurita R,Nakamura Y,Lim KC,Engel JD,Zhou J,Cheng T,Zhu X,Zhu P,Shi L
11-0719-42 was used in Flow cytometry/Cell sorting to demonstrate that interferon-mediated cell cycle control underlies the clinical efficacy of glucocorticoids.
Tue May 10 00:00:00 EDT 2022
The small GTPase RAB10 regulates endosomal recycling of the LDL receptor and transferrin receptor in hepatocytes.
Journal of lipid research
Khan TG,Ginsburg D,Emmer BT
11-0719-42 was used in Flow cytometry/Cell sorting to suggest a model in which RAB10 regulates LDL and transferrin uptake by promoting both slow and rapid recycling routes for their respective receptor proteins.
Mon Aug 01 00:00:00 EDT 2022
Common variants in signaling transcription-factor-binding sites drive phenotypic variability in red blood cell traits.
Nature genetics
Choudhuri A,Trompouki E,Abraham BJ,Colli LM,Kock KH,Mallard W,Yang ML,Vinjamur DS,Ghamari A,Sporrij A,Hoi K,Hummel B,Boatman S,Chan V,Tseng S,Nandakumar SK,Yang S,Lichtig A,Superdock M,Grimes SN,Bowman TV,Zhou Y,Takahashi S,Joehanes R,Cantor AB,Bauer DE,Ganesh SK,Rinn J,Albert PS,Bulyk ML,Chanock SJ,Young RA,Zon LI
11-0719 was used in Flow cytometry/Cell sorting to propose that the majority of the RBC-trait-associated variants that reside on transcription-factor-binding sequences fall in STF target sequences, suggesting that the phenotypic variation of RBC traits could stem from altered responsiveness to extracellular stimuli.
Tue Dec 01 00:00:00 EST 2020
Transcriptional States and Chromatin Accessibility Underlying Human Erythropoiesis.
Cell reports
Ludwig LS,Lareau CA,Bao EL,Nandakumar SK,Muus C,Ulirsch JC,Chowdhary K,Buenrostro JD,Mohandas N,An X,Aryee MJ,Regev A,Sankaran VG
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Tue Jun 11 00:00:00 EDT 2019
Methods for Systematic Identification of Membrane Proteins for Specific Capture of Cancer-Derived Extracellular Vesicles.
Cell reports
Zaborowski MP,Lee K,Na YJ,Sammarco A,Zhang X,Iwanicki M,Cheah PS,Lin HY,Zinter M,Chou CY,Fulci G,Tannous BA,Lai CP,Birrer MJ,Weissleder R,Lee H,Breakefield XO
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Tue Apr 02 00:00:00 EDT 2019
Gene-centric functional dissection of human genetic variation uncovers regulators of hematopoiesis.
eLife
Nandakumar SK,McFarland SK,Mateyka LM,Lareau CA,Ulirsch JC,Ludwig LS,Agarwal G,Engreitz JM,Przychodzen B,McConkey M,Cowley GS,Doench JG,Maciejewski JP,Ebert BL,Root DE,Sankaran VG
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Thu May 09 00:00:00 EDT 2019
GATA/Heme Multi-omics Reveals a Trace Metal-Dependent Cellular Differentiation Mechanism.
Developmental cell
Tanimura N,Liao R,Wilson GM,Dent MR,Cao M,Burstyn JN,Hematti P,Liu X,Zhang Y,Zheng Y,Keles S,Xu J,Coon JJ,Bresnick EH
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Mon Sep 10 00:00:00 EDT 2018
Mechanisms of establishment and functional significance of DNA demethylation during erythroid differentiation.
Blood advances
Bartholdy B,Lajugie J,Yan Z,Zhang S,Mukhopadhyay R,Greally JM,Suzuki M,Bouhassira EE
11-0719 was used in Flow cytometry/Cell sorting to study the mechanisms of establishment and functional significance of DNA demethylation during erythroid differentiation.
Tue Aug 14 00:00:00 EDT 2018
Functional Selectivity in Cytokine Signaling Revealed Through a Pathogenic EPO Mutation.
Cell
Kim AR,Ulirsch JC,Wilmes S,Unal E,Moraga I,Karakukcu M,Yuan D,Kazerounian S,Abdulhay NJ,King DS,Gupta N,Gabriel SB,Lander ES,Patiroglu T,Ozcan A,Ozdemir MA,Garcia KC,Piehler J,Gazda HT,Klein DE,Sankaran VG
11-0719 was used in Flow cytometry/Cell sorting to demonstrate how variation in a single cytokine can lead to biased downstream signaling and can thereby cause human disease.
Thu Mar 09 00:00:00 EST 2017
Variant-aware saturating mutagenesis using multiple Cas9 nucleases identifies regulatory elements at trait-associated loci.
Nature genetics
Canver MC,Lessard S,Pinello L,Wu Y,Ilboudo Y,Stern EN,Needleman AJ,Galactéros F,Brugnara C,Kutlar A,McKenzie C,Reid M,Chen DD,Das PP,A Cole M,Zeng J,Kurita R,Nakamura Y,Yuan GC,Lettre G,Bauer DE,Orkin SH
11-0719 was used in Flow cytometry/Cell sorting to demonstrate the use of multiple designer nucleases and variant-aware library design to interrogate trait-associated regulatory DNA at high resolution.
Sat Apr 01 00:00:00 EDT 2017
Activation of KLF1 Enhances the Differentiation and Maturation of Red Blood Cells from Human Pluripotent Stem Cells.
Stem cells (Dayton, Ohio)
Yang CT,Ma R,Axton RA,Jackson M,Taylor AH,Fidanza A,Marenah L,Frayne J,Mountford JC,Forrester LM
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Sat Apr 01 00:00:00 EDT 2017
Genetically engineered red cells expressing single domain camelid antibodies confer long-term protection against botulinum neurotoxin.
Nature communications
Huang NJ,Pishesha N,Mukherjee J,Zhang S,Deshycka R,Sudaryo V,Dong M,Shoemaker CB,Lodish HF
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Mon Sep 04 00:00:00 EDT 2017
Hormonal vitamin D up-regulates tissue-specific PD-L1 and PD-L2 surface glycoprotein expression in humans but not mice.
The Journal of biological chemistry
Dimitrov V,Bouttier M,Boukhaled G,Salehi-Tabar R,Avramescu RG,Memari B,Hasaj B,Lukacs GL,Krawczyk CM,White JH
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Fri Dec 15 00:00:00 EST 2017
Knockdown of HSPA9 induces TP53-dependent apoptosis in human hematopoietic progenitor cells.
PloS one
Liu T,Krysiak K,Shirai CL,Kim S,Shao J,Ndonwi M,Walter MJ
11071942 was used in flow cytometry to determine that HSPA9 knockdown is associated with increased TP53 expression and activity
Fri Aug 25 00:00:00 EDT 2017
Targeted Application of Human Genetic Variation Can Improve Red Blood Cell Production from Stem Cells.
Cell stem cell
Giani FC,Fiorini C,Wakabayashi A,Ludwig LS,Salem RM,Jobaliya CD,Regan SN,Ulirsch JC,Liang G,Steinberg-Shemer O,Guo MH,Esko T,Tong W,Brugnara C,Hirschhorn JN,Weiss MJ,Zon LI,Chou ST,French DL,Musunuru K,Sankaran VG
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Thu Jan 07 00:00:00 EST 2016
Enforced Expression of HOXB4 in Human Embryonic Stem Cells Enhances the Production of Hematopoietic Progenitors but Has No Effect on the Maturation of Red Blood Cells.
Stem cells translational medicine
Jackson M,Ma R,Taylor AH,Axton RA,Easterbrook J,Kydonaki M,Olivier E,Marenah L,Stanley EG,Elefanty AG,Mountford JC,Forrester LM
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Mon Aug 01 00:00:00 EDT 2016
Influenza Vaccination Generates Cytokine-Induced Memory-like NK Cells: Impact of Human Cytomegalovirus Infection.
Journal of immunology (Baltimore, Md. : 1950)
Goodier MR,Rodriguez-Galan A,Lusa C,Nielsen CM,Darboe A,Moldoveanu AL,White MJ,Behrens R,Riley EM
11-0719 was used in Flow cytometry/Cell sorting to examine the effect of seasonal influenza vaccination on natural killer cell function and phenotype in previously unvaccinated individuals.
Fri Jul 01 00:00:00 EDT 2016
Single-cell profiling of human megakaryocyte-erythroid progenitors identifies distinct megakaryocyte and erythroid differentiation pathways.
Genome biology
Psaila B,Barkas N,Iskander D,Roy A,Anderson S,Ashley N,Caputo VS,Lichtenberg J,Loaiza S,Bodine DM,Karadimitris A,Mead AJ,Roberts I
11-0719 was used in Flow cytometry/Cell sorting to clarify the cellular hierarchy in human megakaryocyte/erythroid lineage commitment and highlight the importance of using a combination of single-cell approaches to dissect cellular heterogeneity.
Tue May 03 00:00:00 EDT 2016
PPAR-α and glucocorticoid receptor synergize to promote erythroid progenitor self-renewal.
Nature
Lee HY,Gao X,Barrasa MI,Li H,Elmes RR,Peters LL,Lodish HF
11-0719 was used in Flow cytometry/Cell sorting to study the role of PPAR-α agonists in stimulating self-renewal of early erythroid progenitor cells.
Thu Jun 25 00:00:00 EDT 2015
BCL11A enhancer dissection by Cas9-mediated in situ saturating mutagenesis.
Nature
Canver MC,Smith EC,Sher F,Pinello L,Sanjana NE,Shalem O,Chen DD,Schupp PG,Vinjamur DS,Garcia SP,Luc S,Kurita R,Nakamura Y,Fujiwara Y,Maeda T,Yuan GC,Zhang F,Orkin SH,Bauer DE
11-0719 was used in Flow cytometry/Cell sorting to develop pooled CRISPR-Cas9 guide RNA libraries to perform in situ saturating mutagenesis of human and mouse enhancers.
Thu Nov 12 00:00:00 EST 2015
Acetylated histones contribute to the immunostimulatory potential of neutrophil extracellular traps in systemic lupus erythematosus.
Clinical and experimental immunology
Pieterse E,Hofstra J,Berden J,Herrmann M,Dieker J,van der Vlag J
11-0719 was used in Flow cytometry/Cell sorting to assess the effect of specific histone modifications on NETs ability to drive autoimmune response in systemic lupus erythematous.
Thu Jan 01 00:00:00 EST 2015
Functional plasticity of the N-methyl-d-aspartate receptor in differentiating human erythroid precursor cells.
American journal of physiology. Cell physiology
Hänggi P,Telezhkin V,Kemp PJ,Schmugge M,Gassmann M,Goede JS,Speer O,Bogdanova A
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Mon Jun 15 00:00:00 EDT 2015
DECKO: Single-oligo, dual-CRISPR deletion of genomic elements including long non-coding RNAs.
BMC genomics
Aparicio-Prat E,Arnan C,Sala I,Bosch N,Guigó R,Johnson R
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Fri Oct 23 00:00:00 EDT 2015
Allele-specific analysis of DNA replication origins in mammalian cells.
Nature communications
Bartholdy B,Mukhopadhyay R,Lajugie J,Aladjem MI,Bouhassira EE
11-0719 was used in Flow cytometry/Cell sorting to delineate the mechanisms that control the location and timing of firing of replication origins.
Tue May 19 00:00:00 EDT 2015
Altered translation of GATA1 in Diamond-Blackfan anemia.
Nature medicine
Ludwig LS,Gazda HT,Eng JC,Eichhorn SW,Thiru P,Ghazvinian R,George TI,Gotlib JR,Beggs AH,Sieff CA,Lodish HF,Lander ES,Sankaran VG
11-0719 was used in Flow cytometry/Cell sorting to study the altered translation of GATA1 in cells isolated from patients with Diamond-Blackfan anemia.
Tue Jul 01 00:00:00 EDT 2014
Terminal differentiation and loss of tumorigenicity of human cancers via pluripotency-based reprogramming.
Oncogene
Zhang X,Cruz FD,Terry M,Remotti F,Matushansky I
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Immunofluorescence
Thu May 02 00:00:00 EDT 2013
A feedback loop consisting of microRNA 23a/27a and the β-like globin suppressors KLF3 and SP1 regulates globin gene expression.
Molecular and cellular biology
Ma Y,Wang B,Jiang F,Wang D,Liu H,Yan Y,Dong H,Wang F,Gong B,Zhu Y,Dong L,Yin H,Zhang Z,Zhao H,Wu Z,Zhang J,Zhou J,Yu J
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Tue Oct 01 00:00:00 EDT 2013
α-1,6-Fucosyltransferase (FUT8) inhibits hemoglobin production during differentiation of murine and K562 human erythroleukemia cells.
The Journal of biological chemistry
Sasaki H,Toda T,Furukawa T,Mawatari Y,Takaesu R,Shimizu M,Wada R,Kato D,Utsugi T,Ohtsu M,Murakami Y
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Fri Jun 07 00:00:00 EDT 2013
Role of ZBP-89 in human globin gene regulation and erythroid differentiation.
Blood
Woo AJ,Kim J,Xu J,Huang H,Cantor AB
Published figure using CD71 (Transferrin Receptor) monoclonal antibody (Product # 11-0719-42) in Flow Cytometry
Thu Sep 29 00:00:00 EDT 2011