Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MPDAFLTCE TVIFAWAIGG EGFSYPPHVG LSLGTPLDPH YVLLEVHYDN PTYEEGLIDN SGLRLFYTMD IRKYDAGVIE AGLWVSLFHT IPPGMPEFQS EGHCTLECLE EALEAEKPSG IHVFAVLLHA HLAGRGIRLR HFRKGKEMKL LAYDDDFDFN FQEFQYLKEE QTILPGDNLI TECRYNTKDR AEMTWGGLST RSEMCLSYLL YYPRINLTRC ASIPDIMEQL QFIGVKEIYR PVTTWPFIIK SPKQYKNLSF MDAMNKFKWT KKEGLSFNEL VLSLPVNVRC SKTDNAEWSI QGMTALPPDI ERPYKAEPLV CGTSSSSSLH RDFSINLLVC LLLLSCTLST KSL (262-613 aa encoded by BC018756)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DBH-like monooxygenase protein 1; DBH-related protein; dopamine-oxygenase; Monooxygenase X; monooxygenase, DBH-like 1
Gene Aliases: 3230402N08Rik; Dbhr; dJ248E1.1; MNCb-5203; MOX; MOXD1; PRO5780; UNQ2493/PRO5780
UniProt ID: (Human) Q6UVY6, (Mouse) Q9CXI3
Entrez Gene ID: (Human) 26002, (Mouse) 59012, (Rat) 294119
Molecular Function: hydroxylase oxidoreductase metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support