Product References
Lysosomal Acid Lipase Is Required for Donor T Cells to Induce Graft-versus-Host Disease.
Cell reports
Nguyen HD,Ticer T,Bastian D,Kuril S,Li H,Du H,Yan C,Yu XZ
12-7052 was used in Flow cytometry/Cell sorting to reveal the role of LAL in T cell alloresponse and pathogenicity and validates LAL as a target for controlling GVHD and tumor relapse after allo-HCT.
Tue Oct 27 00:00:00 EDT 2020
CD4+ T Cells Cross-Reactive with Dengue and Zika Viruses Protect against Zika Virus Infection.
Cell reports
Wen J,Wang YT,Valentine KM,Dos Santos Alves RP,Xu Z,Regla-Nava JA,Ngono AE,Young MP,Ferreira LCS,Shresta S
12-7052 was used in Flow cytometry/Cell sorting to examine the ability of DENV/ZIKV-cross-reactive CD4+ T cells to protect against versus exacerbate ZIKV infection.
Tue Apr 28 00:00:00 EDT 2020
An Id2RFP-Reporter Mouse Redefines Innate Lymphoid Cell Precursor Potentials.
Immunity
Xu W,Cherrier DE,Chea S,Vosshenrich C,Serafini N,Petit M,Liu P,Golub R,Di Santo JP
12-7052 was used in Flow cytometry/Cell sorting to suggest a revised model for ILC differentiation that redefines the cell-fate potential of helper-ILC-restricted Zbtb16+ ILCPs.
Tue Apr 16 00:00:00 EDT 2019
Ovalbumin induces natural killer cells to secrete Th2 cytokines IL‑5 and IL‑13 in a mouse model of asthma.
Molecular medicine reports
Chen Z,Wang L
12-7052-82 was used in Flow Cytometry to analyse the dynamic changes of NK cells and their subsets during the development of the ovalbumin (OVA)‚induced allergic airway response.
Mon Apr 01 00:00:00 EDT 2019
Dephosphorylated Polymerase I and Transcript Release Factor Prevents Allergic Asthma Exacerbations by Limiting IL-33 Release.
Frontiers in immunology
Ni Y,Hao J,Hou X,Du W,Yu Y,Chen T,Wei Z,Li Y,Zhu F,Wang S,Liang R,Li D,Lu Y,Liao K,Li B,Shi G
Mon Mar 21 00:00:00 EDT 2022
Bcl11b is essential for licensing Th2 differentiation during helminth infection and allergic asthma.
Nature communications
Lorentsen KJ,Cho JJ,Luo X,Zuniga AN,Urban JF,Zhou L,Gharaibeh R,Jobin C,Kladde MP,Avram D
12-7052 was used in Flow cytometry/Cell sorting to establish novel roles for Bcl11b in the regulatory loop that licenses Th2 program in vivo.
Thu Apr 26 00:00:00 EDT 2018
Regulatory Innate Lymphoid Cells Control Innate Intestinal Inflammation.
Cell
Wang S,Xia P,Chen Y,Qu Y,Xiong Z,Ye B,Du Y,Tian Y,Yin Z,Xu Z,Fan Z
12-7052 was used in Flow cytometry/Cell sorting to identify a regulatory subpopulation of innate lymphoid cells that exists in the gut, and harbour a unique gene identity.
Thu Sep 21 00:00:00 EDT 2017
Water-soluble chitosan inhibits nerve growth factor and attenuates allergic inflammation in mite allergen-induced allergic rhinitis.
The Journal of allergy and clinical immunology
Chen PC,Hsieh MH,Kuo WS,Kao HF,Hsu CL,Wang JY
12-7052 was used in Flow cytometry/Cell sorting to evaluate the effect of extensive water-soluble chitosan in allergic rhinitis.
Sun Oct 01 00:00:00 EDT 2017
Intestinal helminth infection induces highly functional resident memory CD4+ T cells in mice.
European journal of immunology
Steinfelder S,Rausch S,Michael D,Kühl AA,Hartmann S
Wed Feb 01 00:00:00 EST 2017
Toxoplasma Co-infection Prevents Th2 Differentiation and Leads to a Helminth-Specific Th1 Response.
Frontiers in cellular and infection microbiology
Ahmed N,French T,Rausch S,Kühl A,Hemminger K,Dunay IR,Steinfelder S,Hartmann S
12-7052 was used in Flow cytometry/Cell sorting to conclude a previous T. gondii infection limits a helminth-specific Th2 immune response while promoting a shift toward a Th1-type immune response.
Mon Apr 02 00:00:00 EDT 2018
Lack of autophagy induces steroid-resistant airway inflammation.
The Journal of allergy and clinical immunology
Suzuki Y,Maazi H,Sankaranarayanan I,Lam J,Khoo B,Soroosh P,Barbers RG,James Ou JH,Jung JU,Akbari O
12-7052 was used in Flow cytometry/Cell sorting to examine the role of autophagy in lung inflammation and the pathogenesis of corticosteroid-resistant neutrophilic asthma.
Sun May 01 00:00:00 EDT 2016
Characterization and Quantification of Innate Lymphoid Cell Subsets in Human Lung.
PloS one
De Grove KC,Provoost S,Verhamme FM,Bracke KR,Joos GF,Maes T,Brusselle GG
12-7052 was used in Flow cytometry/Cell sorting to study innate lymphoid cells in the human lung in healthy patients and those with chronic pulmonary disease.
Fri Jul 08 00:00:00 EDT 2016
Transcription Factors Oct-1 and GATA-3 Cooperatively Regulate Th2 Cytokine Gene Expression via the RHS5 within the Th2 Locus Control Region.
PloS one
Kim K,Kim N,Lee GR
12-7052 was used in Flow cytometry/Cell sorting to investigate the molecular mechanisms behind Th2 cytokine gene regulation.
Thu Jul 07 00:00:00 EDT 2016
The IL-10 polarized cytokine pattern in innate and adaptive immunity cells contribute to the development of FVIII inhibitors.
BMC hematology
Silveira AC,Santana MA,Ribeiro IG,Chaves DG,Martins-Filho OA
12-7052 was used in Flow cytometry/Cell sorting to understand immune response to FVIII in patients with HA who were either positive [HAα-FVIII(+)] or negative [HAα-FVIII(-)] for inhibitors.
Thu Feb 05 00:00:00 EST 2015
TNF superfamily member TL1A elicits type 2 innate lymphoid cells at mucosal barriers.
Mucosal immunology
Yu X,Pappu R,Ramirez-Carrozzi V,Ota N,Caplazi P,Zhang J,Yan D,Xu M,Lee WP,Grogan JL
12-7052 was used in an ELISA assay to demonstrate a key role for TL1A in promoting ILC2s at mucosal barriers.
Thu May 01 00:00:00 EDT 2014
Group V secretory phospholipase A2 is involved in macrophage activation and is sufficient for macrophage effector functions in allergic pulmonary inflammation.
Journal of immunology (Baltimore, Md. : 1950)
Ohta S,Imamura M,Xing W,Boyce JA,Balestrieri B
12-7052 was used in Flow cytometry/Cell sorting to identify a unique function for gV-sPLA2 in the activation and recruitment of immune cells.
Sat Jun 15 00:00:00 EDT 2013
Thymic stromal lymphopoietin induces corticosteroid resistance in natural helper cells during airway inflammation.
Nature communications
Kabata H,Moro K,Fukunaga K,Suzuki Y,Miyata J,Masaki K,Betsuyaku T,Koyasu S,Asano K
12-7052 was used in Flow cytometry/Cell sorting to investigate the role of the IL-33/NH-cell axis in corticosteroid resistance.
Thu Jun 12 00:00:00 EDT 2014
Transcription factor RORα is critical for nuocyte development.
Nature immunology
Wong SH,Walker JA,Jolin HE,Drynan LF,Hams E,Camelo A,Barlow JL,Neill DR,Panova V,Koch U,Radtke F,Hardman CS,Hwang YY,Fallon PG,McKenzie AN
12-7052 was used in Flow cytometry/Cell sorting to study how nuocytes arose in the bone marrow and differentiated from common lymphoid progenitors, indicating they are a distinct, previously unknown member of the lymphoid lineage.
Sun Jan 22 00:00:00 EST 2012
Lack of IL-4 receptor expression on T helper cells reduces T helper 2 cell polyfunctionality and confers resistance in allergic bronchopulmonary mycosis.
Mucosal immunology
Müller U,Piehler D,Stenzel W,Köhler G,Frey O,Held J,Grahnert A,Richter T,Eschke M,Kamradt T,Brombacher F,Alber G
12-7052 was used in Flow cytometry/Cell sorting to study the function of the interleukin-4 receptor on T helper cells in pulmonary cryptococcosis.
Tue May 01 00:00:00 EDT 2012
Donor B cells in transplants augment clonal expansion and survival of pathogenic CD4+ T cells that mediate autoimmune-like chronic graft-versus-host disease.
Journal of immunology (Baltimore, Md. : 1950)
Young JS,Wu T,Chen Y,Zhao D,Liu H,Yi T,Johnston H,Racine J,Li X,Wang A,Todorov I,Zeng D
12-7052 was used in Flow cytometry/Cell sorting to investigate the mechanisms whereby donor B cells augment chronic graft-versus-host disease (cGVHD) pathogenesis, showing that B cells augment clonal expansion and survival of pathogenic CD4+ T cells that mediate autoimmune-like cGVHD.
Sun Jul 01 00:00:00 EDT 2012
The acute environment, rather than T cell subset pre-commitment, regulates expression of the human T cell cytokine amphiregulin.
PloS one
Qi Y,Operario DJ,Georas SN,Mosmann TR
12-7052 was used in Magnetic cell separation to collate a detailed analysis of the regulation of Amphiregulin expression by human T cell subsets.
Thu Dec 13 00:00:00 EST 2012
Bronchial epithelial cells produce IL-5: implications for local immune responses in the airways.
Cellular immunology
Wu CA,Peluso JJ,Zhu L,Lingenheld EG,Walker ST,Puddington L
12-7052 was used in Immunofluorescence to investigate how IL-5 production by BEC can impact the microenvironment of the lung, modifying pathological and protective immune responses in the airways.
Wed Sep 01 00:00:00 EDT 2010
Pancreatic lipase-related protein 2 (PLRP2) induction by IL-4 in cytotoxic T lymphocytes (CTLs) and reevaluation of the negative effects of its gene ablation on cytotoxicity.
Journal of leukocyte biology
Alves BN,Leong J,Tamang DL,Elliott V,Edelnant J,Redelman D,Singer CA,Kuhn AR,Miller R,Lowe ME,Hudig D
12-7052 was used in Flow cytometry/Cell sorting to study how extracellular PLRP2 lipase is unable to increase the cytotoxicity lost through PLRP2 ablation, and the overall role of PLRP2 in CD8 T cells..
Tue Sep 01 00:00:00 EDT 2009